Would you like to log in again?
Brady® BradyJet™ J2000 Inkjet Full Color Label Printer, Brady Worldwide
Supplier: Brady Worldwide
J2000-BWSSFID J2000 J2000-BWSLAB
76304-322EA 2945.03 USD InStock
76304-322 76304-320 76304-324
Brady® BradyJet™ J2000 Inkjet Full Color Label Printer, Brady Worldwide
Printers Label Printers
Improve productivity with lab-durable color that survives the harshest lab environments with the BradyJet J2000 color label printer and Brady workstation laboratory identification software suite.
  • Maximum label/tape with 4" width
  • Full color printing capability
  • Ink jet print technology
  • Can be processed maximum 3000 labels per day
  • Maximum print speed of 2.5"/second
  • Easy to change the labels in less than 20 seconds
Quickly and easily differentiate your samples with more than 16 million color options. Easily create the custom labels that fit the needs of your lab with the Brady workstation laboratory identification suite or the safety and facility ID suite software. Protect your research and reduce errors with colors and labels that survive the harshest lab environment so you only have to do your work once. Get performance printing in a small footprint so you can complete identification jobs right, without wasting workspace. Print colors survive liquid nitrogen, DMSO, formalin and more.
Ordering information: Brady J20 labels, tags and accessories are available for these printers.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty