Would you like to log in again?
PlantCon™ Plant Tissue Culture Container, MP Biomedicals
Supplier: MP Biomedicals
092672202
76202-440CS 53.37 USD InStock
76202-440
PlantCon™ Plant Tissue Culture Container, MP Biomedicals
Culture Dishes Cell Culture Dishes
The PlantCon™ container consists of a cover and a bottom and features a vent for gas exchange and a drip control lip to recycle condensation. The cover can be snapped on tight for long-term culture or loosely fitted during acclimation to external conditions prior to transplantation. Provided presterilized, made of clear unbreakable plastic. These disposable space saving containers are specifically designed for optimal plant cell culture applications.
  • Aseptic germination of seeds or spores
  • Stage I production of clean explant for multiplication of callus formation
  • Stage II multiplication of plants through enhanced axillary branching, adventitious buds, embryoids or callus
  • Stage III pretransplant rooting and hardening of plantlets for transfer to soil
  • As a shipping container for pretransplant plantlets
  • PlantCon™ containers may be used with liquid or agar solidified media
  • Can be used for many plant cell culture procedures that currently utilize glass containers
PlantCon™ offers a line of presterilized, disposable, plastic growth containers. This labware has been designed to reduce the time spent preparing an in vitro growth environment for plant cultures and to minimize the risk of contamination.
The PlantCon™ system allows users to start culturing plant cells and plantlets with the following advantages: unbreakable and safe to use, presterilized and easy to use, clear plastic construction offers excellent light transmission, disposable design eliminates messy glass cleanup, space saving design is stackable, units are manufactured under strict quality control procedures, scientifically designed for plant cell culture, special cover positioning utilizes special lugs in the cover lip, allowing the user to snap on the cover tight for handling or loose for free exchange of atmospheres.
They can be used for many plant cell culture procedures that currently utilize glass containers.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty