Would you like to log in again?
Methacrylated Collagen
Supplier: Advanced BioMatrix
5201-1KIT
103701-166EA 425 USD InStock
103701-166
Methacrylated Collagen
Proteins and Peptides
PhotoCol®-IRG is a methacrylated Type I collagen kit. These 3D collagen gels can be prepared at various concentrations and photocrosslinked (365nm) to provide various gel stiffness. Each kit contains 100 mg of methacrylated collagen, irgacure photoinitiator, acetic acid and a neutralization solution.
  • Highly purified
  • Lyophilized to allow you to choose the desired concentration
  • Photocrosslinkable with a wide variety of photoinitiators
100 mg methacrylated collagen, 50 ml of 20 mm acetic acid, 10 ml of neutralization solution, 100 mg of irgacure 2959
Advanced BioMatrix products are recognized for purity, functionality, quality, and lot-to-lot consistency, allowing for reproducibility in research.
Caution: This product is for R and D use only and is not intended for human or other uses. Please consult the Material Safety Data Sheet for information regarding hazards and safe handling practices.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty