Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N,N-Dimethylacetamide 99%, Reagent Grade

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Acetic acid dimethylamideDimethyl Acetamide

D137510-2.5L
BJD137510-2.5LEA 209.69 USD InStock
BJD137510-2.5L
N,N-Dimethylacetamide 99%, Reagent Grade
N,N-Dimethylacetamide


Formula: CH₃CON(CH₃)₂
MW: 87.12 g/mol
Boiling Pt: 11.5 %(V) (99 °C)
Melting Pt: -20 °C
Density: 0.94 g/cm³ (20 °C)
Flash Pt: 64 °C
Storage Temperature: Ambient
MDL Number: MFCD00008686
CAS Number: 127-19-5
EINECS: 204-826-4
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/89/13/2298913.jpg
Order Now
Specification Test Results
Vapor Pressure 2 mmHg ( 25 °C)
Identity (IR) complying
Assay (GC) Min. 98.5 %
Explosion Limit 11.5 %, 160 °F
Autoignition Temperature 914 °F
Refractive Index n20/D 1.437 (lit.)
Vapor Density 3 (vs air)
Explosion Limit 1.8 %, 100 °F

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty