Would you like to log in again?
Disposable Cuvettes, Malvern Panalytical
Supplier: MALVERN PANALYTICAL MS
DTS1070
MSPP-DTS1070PK 407.69 USD InStock
MSPP-DTS1070
Disposable Cuvettes, Malvern Panalytical
Cuvettes Spectrophotometer Cuvettes
These disposable cuvettes are designed primarily for the measurement of Zeta potential with the Zetasizer nano series, but can be used for size measurement.
Ordering information: Requires Zetasizer software version 7.02 or later.
Packaging: Pack of 10 cells with 20 stoppers.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty