Would you like to log in again?
Lentiviral Packaging Plasmid Mix, Cellecta
Supplier: Cellecta
CPCP-K2A
MSPP-CPCPK2AEA 623.08 USD InStock
MSPP-CPCPK2A
Lentiviral Packaging Plasmid Mix, Cellecta
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
Lentiviral packaging plasmid mix allows the production of pseudoviral particles when mixed with second- or third-generation lentiviral vectors.
The packaging plasmid mix is a mixture of the human immunodeficiency virus (HIV) lentiviral packaging plasmid psPAX2 and the pMD2.G plasmid containing VSV-G. These plasmids provide all the necessary structural, regulatory, and replication proteins required to produce pseudotyped packaged lentiviral expression constructs when co-transfected with an HIV-based lentiviral expression construct. Pseudotyped lentiviral particles with VSV-G envelope protein can infect a variety of mammalian or non-mammalian, dividing or non-dividing cells.
Lentiviral packaging plasmid mix contains tested anion-exchange purified packaging plasmids pre-mixed at the proper ratio for optimal production of packaged VSV-g pseudotyped lentiviral particles. This reagent is used in conjunction with HEK 293T cells. The ready-to-use lentiviral packaging plasmid mix is compatible with most commercially-available second- and third-generation lentiviral vectors.
Delivery information: Kit contains 250 µg of lentiviral packaging plasmid mix that will be sufficient for 25 transfections in 10 cm culture plates.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty