Would you like to log in again?
MOPS, Free Acid, Molecular Biology Grade, EMD Millipore
Supplier: MilliporeSigma
Calbiochem®
475922-100GM
80503-004EA 125.98 USD InStock
80503-004
MOPS, Free Acid, Molecular Biology Grade, EMD Millipore
Buffers Electrophoresis Buffers Running Buffers
MOPS, free acid is a zwitterionic buffer useful in the pH range of 6.5 to 7.9. Has a pKa of 7.20 at 25 °C.
  • Purity: ≥99% (Potentiometry)
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty