Would you like to log in again?
DirectPCR® Lysis Reagent (Cell), Viagen
Supplier: Viagen
DirectPCR®
302-C
MSPP-302CEA 244.62 USD InStock
MSPP-302C
DirectPCR® Lysis Reagent (Cell), Viagen
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
DirectPCR® DNA extraction system is a single-tube system for rapid preparation of DNA from mouse tails, ear pieces, yolk sac, and culture cells.
  • Time saving: Less hands-on time, crude tail lysates for PCR
  • Safely contains no organic reagents
  • Less waste (organic reagents, tubes, tips)
  • Reliable and efficient with high yields
  • For use with cultured cells
  • Stable up to 12 months at 4 °C
  • PCR genotyping with 1 μl lysates
Crude extracts of biological samples are not compatible with many molecular biology-grade reactions such as polymerase chain reaction (PCR), in part due to inhibitors contained in crude extracts. The DirectPCR reagents mediate the rapid lysis of biological samples and contain inhibitors that effectively suppress the inhibitory activities of crude lysates for PCR amplification, while maximally maintaining the integrity of released genomic DNA. Patent-pending procedure completely eliminate any solution transfer or tube-opening steps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty