Would you like to log in again?
Microcentrifuge Tubes, Forensic DNA Grade, EPPENDORF
Supplier: Eppendorf
0030123611 0030123603 0030122402 0030122259 0030122267 0030119606
76019-018CS 117.87 USD InStock
76019-018 76019-022 76019-012 76019-024 76019-014 76019-020
Microcentrifuge Tubes, Forensic DNA Grade, EPPENDORF
The forensic DNA grade microcentrifuge tubes are used for DNA extraction, sample processing and PCR setup as well as for sample storage.
  • High degree of automation to minimise direct contact between user and tubes
  • Strict adherence to cleaning procedures and protocols minimise the risk of contamination
  • Positive air pressure prevents intrusion of dust
Certifications: Compliance to ISO 18385.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty