Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
ComfortGEL Cleanroom ESD Anti-Fatigue Floor Mats, Transforming Technologies
Supplier: Transforming Technologies
FM62436BK FM62036BK FM66036BK
76209-900EA 459.89 USD InStock
76209-900 76209-896 76209-898
ComfortGEL Cleanroom ESD Anti-Fatigue Floor Mats, Transforming Technologies
Floor Mats Traction and Anti-Fatigue Floor Mats
ComfortGEL ESD Anti-Fatigue Mats prevent physical discomfort, stress, and fatigue in controlled environments.
  • Made with rubberized gel foam for maximum softness and cushioning
  • Non-slip, water resistant design
  • Exceptional comfort and long-term performance
  • Static dissipative
  • Specially formulated antimicrobial and antibacterial material to promote cleanliness
This non-slip, water resistant mat is constructed from a close cell rubberized gel foam which conforms to feet while promoting proper circulation and providing maximum comfort. Mats are available in three sizes, and are finished with beveled edges. They can be cleaned to cleanroom level100.
Certifications: Meets or exceeds requirements of ANSI ESD-S20.20 and the recommendations of ESD 4.1.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty