Would you like to log in again?
Centrifuge Tubes, Unthreaded, Caplugs
Supplier: Caplugs
220-3937-080 214-2415-01K 300-3541-G20
80700-710CS 454.37 USD InStock
80700-710 80700-714 80700-780
Centrifuge Tubes, Unthreaded, Caplugs
Tubes Centrifuge Tubes
Caplugs 15 ml unthreaded centrifuge tube is precision molded from premium-grade, chemically-inert thermoplastics. 50 ml centrifuge tubes are ideal for general purpose laboratory work.
  • 50 ml tubes feature molded-in graduations at 5 ml intervals
Accurate, easy-to-read graduations are molded-in at 0.1 ml intervals from 0 to 1 ml and at 0.5 ml intervals from 1 to 5 ml and every 1 ml from 5 to 15 ml.
Polystyrene tubes withstand up to 6200 ×g and are appropriate for use with mild acidic (except oxidizing acids) or alkali solutions, alcohols (except higher alcohols) and salt.
Ordering information: Stopper caps available separately. Available in conical bottom only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty