Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

SAF-T-PAK® Category A Ambient Shipping System

Supplier: SAF-T-PAK INC MS
STP-130
MSPP-STP-130CS 328.48 USD InStock
MSPP-STP-130
SAF-T-PAK® Category A Ambient Shipping System
Mailers and Shippers
Most economical Category A shipper for max. four 10 ml tubes.

System components:

Outer box – OD 11.25 x 7.75 x 7.75”, preprinted with: Class 6 Infectious Substance Label & UN2814 Marking
Inner box – OD 6.75 x 3.25 x 3.5”
STP-150 50 mL absorbent strip (1 ea.)
STP-609 6 x 12” Saf-T-Rap® bubble wrap (1 ea.)
STP-700 Disposable 2-part secondary pressure vessel, small (1 ea.)
STP-810 Blockout sticker (1 ea.)

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty