Would you like to log in again?
G-Rex® 6M Well Plate, Wilson Wolf
Supplier: WILSON WOLF MANUFACTURING CORP MS
80660M
MSPP-80660MEA 582.65 USD InStock
MSPP-80660M
G-Rex® 6M Well Plate, Wilson Wolf
Microplates Deep Well Microplates
Individual well specifications of 10 cm² gas permeable membrane. Expand 5 million cells into between 200 to 400 million cells in about 10 days with no medium exchange.
  • Gamma irrradiated
Caution: For non-clinical, non-therapeutic, research use only.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty