Would you like to log in again?
Centrifuge Tube with Short Conical Bottom, Chemglass
Supplier: Chemglass
CG-4013-01
80072-368EA 59.23 USD InStock
80072-368
Centrifuge Tube with Short Conical Bottom, Chemglass
Tubes Centrifuge Tubes
These tubes are supplied complete with a poly-seal polypropylene-lined phenolic screw cap. Centrifuge tube with a short, conical bottom and 24-415 GPI threaded top. Fits I.E.C. shield number 320.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty