Would you like to log in again?
Fetal Bovine Serum, U.S. Origin, Genetex
Supplier: GeneTex
GTX73252
89347-790EA 1305.58 USD InStock
89347-790
Fetal Bovine Serum, U.S. Origin, Genetex
Sera (Serum)
FBS is ideal for use in ELISA, western blotting, immunohistochemistry, immunofluoroscence and other applications.
The serum has to be stored at –80 °C or below. Freeze-thaw cycles are to be avoided and standard sterile procedures are to be handled to maintain it's sterility.
Caution: For In vitro laboratory use only. Not for any clinical, therapeutic, or diagnostic use in humans or animals. Not for animal or human consumption.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty