Would you like to log in again?
Spore Strips, CROSSTEX MS
Supplier: Crosstex
DS-500 DS-1000 DS-100
101101-832PK 843.08 USD InStock
101101-832 101101-830 101101-828
Spore Strips, CROSSTEX MS
Sterilization Indicators
Geobacillus stearothermophilus and B.atrophaeus dual species spore strips are developed specifically for direct inoculation of products and for the preparation of biological indicators.
  • Two years shelf life from date of manufacture
  • All concentrations are per ml and contain 20% ethanol
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty