Would you like to log in again?
SpeedMixer™ Disposable Jars, 150 Series, Flacktek
Supplier: FlackTek
501221P-J
MSPP-501221PJCS 463.02 USD InStock
MSPP-501221PJ
SpeedMixer™ Disposable Jars, 150 Series, Flacktek
Jars General Use Jars
These disposable jars eliminate clean up and have the added advantage of enabling the user to store material once mixed.
  • Minimize vibrations and reduce excessive wear on the machine
  • White color
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty