Would you like to log in again?
Chloride Test Kit, LaMotte®
Supplier: LaMotte
R-4503-DR-01 4503-DR-02
470308-450EA 73.15 USD InStock
470308-450 470308-452
Chloride Test Kit, LaMotte®
Environmental Test Kits
This kit tests for Chloride in fresh water samples using the Direct Reading Titrator method.
  • Range 0-200 ppm
  • 50 Tests
Includes 4 reagents.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty