Would you like to log in again?
Debakey Pediatric Forceps, Sklar
Supplier: Sklar
52-1904
10199-370EA 497.28 USD InStock
10199-370
Debakey Pediatric Forceps, Sklar
Forceps
Designed for use on pediatric patients. Forceps are commonly used for grasping fine tissue and blood vessels or soft tissue dissection. The narrow tips and atraumatic teeth cause little to no damage to the tissues.
  • Nontraumatic fine teeth: 1×2 rows
  • Finger ring handles
  • Reusable
  • Latex-free
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty