Would you like to log in again?
Weingartner Ear Forceps, Sklar
Supplier: Sklar
67-8155
10198-480EA 755.59 USD InStock
10198-480
Weingartner Ear Forceps, Sklar
Forceps
Alligator style forceps with serrated jaw to minimize slippage. Useful in middle ear procedures where the powerful grasp and curved jaws assist in the removal of foreign objects.
  • Finger ring handles
  • Non locking
  • Reusable
  • Latex-free
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty