Would you like to log in again?
German Glass Coverslips, PDL Coated, Neuvitro Corporation
Supplier: NEUVITRO CORPORATION MS
GG-15-PDL
MSPP-GG15PDLCS 128.58 USD InStock
MSPP-GG15PDL
German Glass Coverslips, PDL Coated, Neuvitro Corporation
Cover Glasses
These are crystal clear German glass coverslips with poly-D-lysine coating on both sides mainly used for cell culture and confocal fluorescence microscopy.
  • Sterilized
  • Ready to use
Certifications: CE certified.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty