Would you like to log in again?
Native Bovine Fibronectin (from Plasma), Calbiochem®
Supplier: MilliporeSigma
341631-1MG
80051-634EA 224.77 USD InStock
80051-634
Native Bovine Fibronectin (from Plasma), Calbiochem®
Proteins and Peptides
Native fibronectin is purified from pooled bovine plasma. Effective agent for promoting attachment of cells to commonly used culture substrates.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty