Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(-)-Cystine ≥98%, Calbiochem®

Supplier: MilliporeSigma
Synonyms: (2R,2'R)-3,3'-Disulfanediylbis(2-aminopropanoic acid)(2R,2'R)-3,3'-Disulphanediylbis(2-aminopropanoic acid)(H-Cys-OH)2(R)-(-)-Cystine

2470-25GM 2470-25GM
80057-978EA 84.49 USD InStock
CA80057-978 80057-978
L(-)-Cystine ≥98%, Calbiochem®
L(-)-Cystine
Optical rotation: -218° to -224°(c = 2, 1 N HCl, 20°C)


Formula: HOOCCH(NH₂)CH₂SSCH₂CH(NH₂)COOH
MW: 240.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00064228
CAS Number: 56-89-3
Order Now
Specification Test Results
Purity (Macro-Kjeldahl Method) ≥98%

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty