Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Tuberculin Syringe, B. Braun
Supplier: B. Braun
9161502SUS
75796-282CS 790.82 USD InStock
75796-282
Tuberculin Syringe, B. Braun
Syringes Syringes, Medical
Syringe with consistent, dependable performance in a wide range of options.
  • Double seal stopper to prevent medication leakage
  • Large, stable finger grips
  • DEHP-free and latex-free
Smooth plunger action, high visibility - transparent barrel with bold graduations, secure backstop to prevent accidental plunger withdrawal.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty