Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Category B Frozen Shipper, 95 kPa Canister, Therapak®

Supplier: Therapak
380134G
76203-888CS 363.59 USD InStock
76203-888
Category B Frozen Shipper, 95 kPa Canister, Therapak®
Mailers and Shippers
Frozen shipping system, category B with 95kPa canister.

Delivery information: The system includes: transport box (OD: 11.5×9.5×8"), compact cooler (ID: 8×6×4"), 95 kPa canister, 6-bay Aqui-Pak absorbent pouch, cushioning material, list of contents card, dry ice label, and required labels for IATA PI650 marking and compliance.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty