Would you like to log in again?
Reflux Condensers, Coil Type, Chemglass
Supplier: Chemglass
CG-1216-03 CG-1216-05 CG-1216-01
80067-352EA 414.25 USD InStock
80067-352 80067-356 80067-348
Reflux Condensers, Coil Type, Chemglass
Condensers
Reflux condenser with the coolant circulated through the inner coil instead of through the jacket as in the graham style condenser.
  • Condenser has standard taper top outer and lower inner drip tip joints
  • Hose connections have an O.D. of 10 mm at the middle serration
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty