Would you like to log in again?
VH₂O₂ Chemical Indicator Strips, Crosstex®
Supplier: Crosstex
GPS-250-R
101101-688PK 28 USD InStock
101101-688
VH₂O₂ Chemical Indicator Strips, Crosstex®
Test Paper Indicator Paper
This is intended for use as an internal chemical indicator for all packages to be sterilized in hydrogen peroxide sterilization processes.
Provides distinct color change, turning blue when exposed to H₂O₂.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty