Would you like to log in again?
EK-i Series Compact Balance, A and D Engineering
Supplier: A & D ENGINEERING, INC. MS
EK-200I
MSPP-EK200IEA 625 USD InStock
MSPP-EK200I
EK-i Series Compact Balance, A and D Engineering
Balances and Scales
This balance features a hygienic and chemically resistant stainless-steel weighing pan and external calibration.
  • Security ring to prevent theft
  • Resolution of 0.01 g
  • Auto power off function
  • Comparator function with buzzer
Adjustable response characteristics to help cope with the effect of drafts and vibrations.
Multiple weighing units: g, oz, lb, ozt, ct, mom, dwt, gr.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty