Would you like to log in again?
96 well Glass Bottom Plate, Cellvis
Supplier: CELLVIS, LLC MS
P96-1.5H-N
MSPP-P9615HNCS 383.08 USD InStock
MSPP-P9615HN
96 well Glass Bottom Plate, Cellvis
Microplates Cell Culture Plates
Black polystyrene frame with high performance cover glass (0.170 ±0.005 mm), with lid, Individually packed.
  • Suitable for long term tissue culture
  • Manufactured in a class 100000 clean room
  • Plate made from virgin polystyrene, tissue culture treated
  • German high quality cover glass of superior optical quality, the cover glass has a thickness of 0.170 ±0.005 mm
  • Sterilized by gamma radiation
Suitable for Differential interference contrast (DIC), widefield fluorescence, confocal microscopy, two-photon and multiphoton microscopy, fluorescence recovery after photobleaching (FRAP), förster resonance energy transfer (FRET), fluorescence lifetime imaging microscopy (FLIM), total internal reflection fluorescence (TIRF) and super-resolution microscopy.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty