Would you like to log in again?
Zirconium Crucibles with Straight Wall, Metal Technology
Supplier: Metal Technology
10-0045SW
80700-504EA 285.96 USD InStock
80700-504
Zirconium Crucibles with Straight Wall, Metal Technology
Crucibles
Zirconium is the most effective crucible material for fusions using sodium carbonate or sodium peroxide and based on an average of 100 fusions per crucible, it is more cost effective than less expensive porcelain or steel crucibles.
  • Low-cost replacement for platinum
  • Resistant to solvents of all concentrations such as nitric, hydrochloric, sulfuric and perchloric acids
The resistant to solvents allows the fusions to be dissolved out of the crucible completely with little or no contamination of the sample. Alkaline solvents such as ammonium hydroxide, acetate sulfide, carbonate and (Na, K) hydroxide, sulfide, carbonate and cyanide are also without effect.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty