Would you like to log in again?
Ward's® Formaldehyde-Free Bullfrogs
Supplier: VWR
Ward's®
WARD470000-726
470000-726PK 179.95 USD InStock
470000-726
Ward's® Formaldehyde-Free Bullfrogs
Specimens Preserved Specimens Preserved Animal Materials
Formaldehyde-free bullfrogs. Available in pails or vacuum packs.
  • Specimens can be used comfortably over extended periods of time
To better serve the needs of all teaching environments and preferences, WARD’S® offers this line as an alternative to our existing line of high-quality, safe preserved specimens. Absolutely no formaldehyde is used in the fixing and preserving process for these specimens.
Instead, this solution used for preparation and binds with the tissue as non-toxic, cross-linked proteins, making the solution truly 100% formaldehyde-free. WARD’S® offers the largest selection of specimens prepared with this alternative process, including the widest range of sizes and injection styles.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty