Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-AGT Rabbit Polyclonal Antibody

Supplier: BioVision
AB286174-100UG
76305-158EA 896.32 USD InStock
76305-158
Anti-AGT Rabbit Polyclonal Antibody
Antibodies
The octapeptide angiotensin II exerts a wide range of effects on the cardiovascular system. It is also implicated in the regulation of cell proliferation, fibrosis and apoptosis. Angiotensin II is formed by cleavage of angiotensin I by angiotensin-converting enzyme (ACE) or chymases. Human heart chymase, a chymotrypsin-like serine proteinase, hydrolyzes the Phe8-His9 bond to yield the octapeptide hormone angiotensin II and His-Leu.

WB: 1-4 µg

Type: Primary
Antigen: AGT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty