Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-HMGA2 Rabbit Polyclonal Antibody

Supplier: BioVision
AB273014-100UL
76305-252EA 1003.58 USD InStock
76305-252
Anti-HMGA2 Rabbit Polyclonal Antibody
Antibodies
Functions as a transcriptional regulator. Functions in cell cycle regulation through CCNA2.

WB 1:500-1:1000, ELISA (peptide) 1:20000-1:40000

Type: Primary
Antigen: HMGA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty