Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dried Cranberries, Sweetened

Supplier: Bio-Serv
F7457 F7457
76285-268BG 11.41 USD InStock
CA76285-268 76285-268
Dried Cranberries, Sweetened
Foodservice Supplies
Sweetened dried cranberries which is 100% natural human food grade product. Excellent reward treat, appetite stimulant or addition to any enrichment program. Shelf life: 12 months, cool dry storage.

  • Protein: 0.0%
  • Fat: 1.0%
  • Fiber: 6.0%
  • Ash: 0.2%
  • Moisture: <16%
  • Carbohydrate: 82.5%
  • Calories: 3.4 kcal/gm

Ingredients: cranberries, sugar, citric acid, sunflower oil, elderberry juice concentrate.

Packaging: Conveniently packaged with a long shelf life.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty