Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dexpramipexole dihydrochloride ≥98%

Warning
Synonyms: (R)-Pramipexole dihydrochloride(R)-N6-propyl-4,5,6,7-tetrahydrobenzo[d]thiazole-2,6-diamine dihydrochloride

AB254233-25MG AB254233-5MG
76303-384EA 206.84 USD InStock
76303-384 76303-386
Dexpramipexole dihydrochloride ≥98%
Dexpramipexole dihydrochloride
Dexpramipexole dihydrochloride is a neuroprotectant that may slow the progression of ALS by keeping the power on in deteriorating motor nerves. Dexpramipexole maintains mitochondrial function in people with ALS by increasing the efficiency of ATP production.

  • A neuroprotectant


Formula: C₁₀H₁₇N₃S.2HCl
MW: 284.2 g/mol
Storage Temperature: Freezer
MDL Number: MFCD13186799
CAS Number: 104632-27-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty