Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

TP-3654 ≥98%

Supplier: BioVision
Danger
Synonyms: 2-(trans-4-((3-(3-(Trifluoromethyl)phenyl)imidazo[1,2-b]pyridazin-6-yl)amino)cyclohexyl)propan-2-ol2-((1R,4R)-4-((3-(3-(trifluoromethyl)phenyl)imidazo[1,2-b]pyridazin-6-yl)amino)cyclohexyl)propan-2-ol

AB286959-25MG AB286959-5MG
76303-432EA 245.13 USD InStock
76303-432 76303-434
TP-3654 ≥98%
TP-3654
TP-3654 is a second-generation Pim kinase inhibitor with Ki values of 5 and 42 nM for Pim-1 and Pim-3, respectively. Displays anticancer activities.

  • A potent second-generation Pim kinase inhibitor


Formula: C₂₂H2₂₅F₃N₄O
MW: 418.46 g/mol
Storage Temperature: Freezer
MDL Number: MFCD28502206
CAS Number: 1361951-15-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty