Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

MOZ-IN-3 ≥98%

Supplier: BioVision
Synonyms: N'-(benzenesulfonyl)-2-fluoro-3-methyl-5-phenylbenzohydrazideN'-(benzenesulphonyl)-2-fluoro-3-methyl-5-phenylbenzohydrazide

AB286965-5MG AB286965-25MG
76303-458EA 1225.73 USD InStock
76303-458 76303-558
MOZ-IN-3 ≥98%
MOZ-IN-3
MOZ-IN-3 is a potent inhibitor of MOZ, a member of histone acetyltransferases (IC₅₀ = 55 nM).

  • A potent MOZ inhibitor


Formula: C₂₀H₁₇FN₂O₃S
MW: 384.42 g/mol
Storage Temperature: Freezer
CAS Number: 2055397-18-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty