Would you like to log in again?
TCH® Defined Serum Replacement, 50X Concentrate, MP Biomedical,
Supplier: MP Biomedicals
092020026
IC092020026EA 279.05 USD InStock
IC092020026
TCH® Defined Serum Replacement, 50X Concentrate, MP Biomedical,
Cell Culture Media
This serum replacement is effective in research situations such as studying the effects of growth factors or in the manufacturing of cell derived products.
  • A universal serum replacement that supports human and mammalian cells
  • Completely free of virus, mycoplasma and other infectious agents
  • Contains no growth factors, steroid hormones, extraneous proteins or phenol red
  • Protein origin - human; Drug Master on file with the FDA
TCH is a fortified, multipurpose serum replacement for long-term culturing of primarily human anchorage-dependent and suspension cells. Highly effective in the production of biologicals, as well as in culturing bacteria cell culture and other mammalian cells.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty