Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Alsever's Solution, MP Biomedicals
Supplier: MP Biomedicals
092801154
IC092801154EA 75.85 USD InStock
IC092801154
Alsever's Solution, MP Biomedicals
Cell Culture Media
Alsever's Solution is routinely used as an anti-coagulant/blood preservative, which permits the storage of whole blood at refrigerator temperatures for approximately 10 weeks.
  • Isotonic, balanced salt solution
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty