Would you like to log in again?
Human Serum, Pooled (HCV, HIV, HBsAG Free), MP Biomedical, LLC.
Supplier: MP Biomedicals
092931149
IC092931149EA 343.86 USD InStock
IC092931149
Human Serum, Pooled (HCV, HIV, HBsAG Free), MP Biomedical, LLC.
Sera (Serum)
Human Serum, Pooled (HCV, HIV, HBsAG Free) is a base matrix for hormone and steroid calibrators, controls, standards and linearity products.
  • Obtained from normal, clotted blood
  • Liquid media
  • Store at —20 °C
It is also found to have hormone and steroid mass spectrometry applications.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty