Would you like to log in again?
Goat Serum, MP Biomedical, LLC.
Supplier: MP Biomedicals
092939149 092939154
IC092939149EA 98.51 USD InStock
IC092939149 IC092939154
Goat Serum, MP Biomedical, LLC.
Sera (Serum)
Goat serum is a suitable alternative to fetal bovine serum in a wide range of cell culture applications such as the isolation and identification of viruses produced in a variety of primary cell lines.
  • Produced from whole blood collected from healthy adult animals
  • Liquid Media
  • Store at —20 °C
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty