Would you like to log in again?
Absorption Triangles, Fine Science Tools
Supplier: Fine Science Tools
18105-03
MSPP-1810503PK 213.85 USD InStock
MSPP-1810503
Absorption Triangles, Fine Science Tools
Wipes Sponges
These synthetic surgical sponges absorb fluids twice as fast as the best cotton sponges.
  • Have three times more fluid-holding capacity
  • Made from strong, soft, lint-free, biocompatible-proven material
The sponges are non-irritating to tissue and leaves no fibrous residue which might cause wound contamination.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty