Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Steel Wheel for Gas Cylinder Hand Trucks, 20", Justrite®

Supplier: Justrite
35392
76292-426EA 230.38 USD InStock
76292-426
Steel Wheel for Gas Cylinder Hand Trucks, 20", Justrite®
Carts
Steel Wheel for Gas Cylinder Hand Trucks provides safety when moving and storing of gas cylinders.

  • Black powder-coated finish
  • All welded 20" steel replacement wheel for double cylinder Justrite® hand trucks
  • Overall size is 20×1.5"

Features 0.5" steel spokes and steel hub with dual ball bearings, sized for 0.625" axle (not included).

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty