Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Semi-Pneumatic Wheel for Gas Cylinder Hand Trucks, 8", Justrite®

Supplier: Justrite
35370
76292-448EA 47.6 USD InStock
76292-448
Semi-Pneumatic Wheel for Gas Cylinder Hand Trucks, 8", Justrite®
Carts
The Semi-Pneumatic Wheel for Gas Cylinder Hand Trucks provides safety when moving and storing of gas cylinders.

  • Semi-Pneumatic 8" replacement wheel for single gas-cylinder Justrite® hand trucks
  • Wheel size is 8×1.75"

Features a welded steel hub with dual ball bearings, sized for a 0.5" axle (not included).

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty