Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Flat-Free Wheel for Gas Cylinder Hand Trucks, 10.5", Justrite®

Supplier: Justrite
35378
76292-456EA 100.1 USD InStock
76292-456
Flat-Free Wheel for Gas Cylinder Hand Trucks, 10.5", Justrite®
Carts
This Flat-Free Wheel for Gas Cylinder Hand Trucks provides safety when moving and storing of gas cylinders.

  • Flat-free 10.5" replacement wheel for single and double cylinder Justrite® hand trucks
  • Urethane filled wheels will not go flat to ensure mobility on almost any terrain

Features welded steel hub with dual ball bearings, sized for 0.625" axle (not included). Size is 10.5×4".

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty