Would you like to log in again?
Histamine Assay Kit (Fluorometric), BioVision, Inc.
Supplier: BioVision
AB273277-100TEST
76305-278EA 929.27 USD InStock
76305-278
Histamine Assay Kit (Fluorometric), BioVision, Inc.
Assays Enzyme Assays
Histamine assay kit is a sensitive enzymatic assay kit for the detection of histamine in biological samples.
  • Simple, rapid and convenient assay
  • Limit of detection: 10 ppb
  • Very specific, does not interact with other bioamines
Histamine is a biogenic amine with considerable biological relevance. As a signaling molecule, histamine plays manifold roles in the body, ranging from local immune responses to neurotransmission. Histamine is also frequently encountered in food, particularly fish and fermented food products such as sauerkraut and aged cheeses.
This is because some bacteria generate histamine from histidine via the enzyme histidine decarboxylase. Elevated levels of bacterial fermentation can lead to elevated histamine in raw meat and food products. Histamine levels thus can be used as an indicator of spoilage. Concentrations of histamine in the human blood are typically in the low nm range and only in instances of acute histamine poisoning would this metabolite be elevated above the sub-micromolar range.
Histamine assay kit can be used to identify low levels of histamine in various fish and products and sauces/beverages, as well as some biological samples. With our kit, as little as ten pmoles of histamine can be detected in samples, or roughly ten parts per billion (ppb).
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty