Would you like to log in again?
PicoProbe™ Fructose-1,6-Bisphosphatase Inhibitor Screening Kit (F), BioVision, Inc.
Supplier: BioVision
AB283382-100TEST
76305-280EA 1149.12 USD InStock
76305-280
PicoProbe™ Fructose-1,6-Bisphosphatase Inhibitor Screening Kit (F), BioVision, Inc.
Assays Protein Assays
Simple assay for high-throughput screening of FBP1 inhibitors/activators.
  • High-throughput compatible
  • Includes inhibitor control (AMP)
Fructose-1,6-bispohphatase (FBP1) is responsible for the conversion of fructose-1,6-bisphosphate to fructose-6-phosphate in the rate determining step of gluconeogenesis. FBP1 is a tetramer that plays a role in regulating adiposity and appetite. FBP1 has emerged as an important therapeutic target to treat diabetes type 2 to regulate glucose levels during gluconeogenesis.
FBP1 inhibitor screening kit, human FBP1 hydrolyzes fructose-1,6-bisphosphate to fructose-6-phosphate which oxidizes and reacts with a fluorogenic probe, that is detected at Ex/Em= 535/587 nm. The assay is fast, sensitive, high-throughput compatible and provides a quick and sensitive way for screening, studying and characterizing potential novel inhibitors of FBP1.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty