Would you like to log in again?
EZClick™ Global RNA Synthesis Assay Kit (FACS/Microscopy), Red Fluorescence, BioVision, Inc.
Supplier: BioVision
AB273285-100TEST
76305-294EA 926.98 USD InStock
76305-294
EZClick™ Global RNA Synthesis Assay Kit (FACS/Microscopy), Red Fluorescence, BioVision, Inc.
Assays Nucleic Acid Assays
Microplate-based assay is used to detect total RNA content using metabolic labeling techniques.
  • Simple and fast
  • Does not require lengthy incubation times
RNA plays crucial role in coding, decoding and regulation of genes, and protein expression in all living cells. The ability to detect newly synthesized RNA or changes in RNA levels under various physiological conditions, or resulting from disease, environmental damage, or drug treatments is an important aspect of toxicological profiling. Many anti-cancer drugs inhibit transcription, and most transcription inhibitors have useful pharmacological properties. BioVision’s EZClick™ global RNA synthesis assay kit provides a simple and robust tool for detection of global RNA transcription, temporally and spatially, or changes in RNA levels directly in living cells.
De novo synthesized RNA can be detected with a simple procedure without the use of radiolabeling or antibodies. The kit principle relies on the incorporation of cell permeable 5-EU (ethynyl uridine) into nascent RNA, but not into DNA, instead of its natural uridine analog. 5-EU can be used as a replacement for BrU (5-bromo-uridine) to measure de novo synthesized RNA in proliferating cells. Modified RNA is detected by click chemistry using an azide-containing dye that enables for multiplex analyses with other probes, or detection of RNA-interactive proteins for deeper biological insights.
The kit provides sufficient materials for 100 assays for quantitative analysis by microplate reader (Ex/Em= 494/521 nm) and or by fluorescence microscope
Delivery information: Includes actinomycin D, an inhibitor RNA synthesis that serves as an experimental control.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty