Would you like to log in again?
Acid Phosphatase Cell Cytotoxicity Assay Kit, BioVision
Supplier: BioVision
AB273300-500TEST
76305-310EA 620.35 USD InStock
76305-310
Acid Phosphatase Cell Cytotoxicity Assay Kit, BioVision
Assays Cellular Assays
Highly sensitive assay to measure cell cytotoxicity.
  • The assay detects acid phosphatase (AP) activity as a direct correlation of cell cytotoxicity
The measurement of the lysosomal acid phosphatase (ACP) enzymatic activity has become one of the common methods used to detect and quantify cell viability or drug cytotoxicity due to ubiquitous of this enzymatic activity among all organisms. Under normal conditions, eukaryotic cells exhibit abundant cell-membrane associated ACP activity; however ACP activity is considerably lower when cells are exposed to cytotoxic agents.
Measurement of the ACP activity in cells is a reliable method to quantify cell growth in both adherent and non-adherent cells. Compared with other cell proliferatioin assays based on the reduction of tetrazolium salts, ACP cytotoxicity assay displays higher sensitivity and reproducibility. In this cell-based assay, intracellular ACP cleaves the colorless chromogenic substrate para-nitrophenyl phosphate producing a yellow colored product (p-nitrophenol) under alkaline conditions.
The released chromophore can be measured at OØ: 405 nm and is directly proportional to the viable cell number. BioVision’s ACP cytotoxicity assay kit is simple, accurate, reproducible and sensitive with a dynamic range between 103 to 105 cells.
This kit offers a simple, yet excellent and efficient method for in vitro cytotoxicity studies as well as high-throughput drug screening.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty