Would you like to log in again?
Protein A Spin Antibody Purification Kit, BioVision
Supplier: BioVision
AB288107-10TEST
76305-312EA 474.07 USD InStock
76305-312
Protein A Spin Antibody Purification Kit, BioVision
Protein Purification Protein Purification Kits
Kit can be used to purify antibodies in serum, ascites and cell culture medium from various species.
  • Ready-to-use pre-packed spin columns of 0.1 ml bead volume in 20% ethanol
  • High binding capacity: Binding of IgG > –30 mg human, mouse or rabbit IgG mg/ml Protein A-Agarose
Protein A beads have been widely used for IgG purification for their ability to bind immunoglobulins. BioVision’s Protein A (Cat. No. 6500) is a genetically engineered protein containing five IgG-binding regions of native protein A.
Maximum IgG specificity is achieved by eliminating its cell wall binding region, albumin binding region, and other non-specific regions.
Hi-bindTM protein A-agarose beads (Cat. No. 6520) for purification of IgG contain covalently coupled recombinant protein A to 6% cross-linked agarose beads, the most popular resin for protein purification. BioVision’s protein A mini antibody purification kit is a simple, ready to use kit containing all necessary buffers and pre-packed columns to purify up to 30 mg/ml of IgG.
The kit can be used to purify antibodies in serum, ascites and cell culture medium from various species such as human, mouse, rat, goat and rabbit.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty